Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                CD34 Antibody (756525) - Azide and BSA Free, R&D Systems™
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | CD34 | 
|---|---|
| Clone | 756525 | 
| Concentration | 1.0 mg/mL | 
| Dilution | Flow Cytometry, CyTOF-ready | 
| Applications | Flow Cytometry, CyTOF | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
                
                
                    
                        MAB72272SP
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        Novus Biologicals
                         MAB72272SP  | 
                
            
            
            
            
            
                
                    25 μg | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $206.50
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
                
                
                    
                        MAB72272
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        Novus Biologicals
                         MAB72272  | 
                
            
            
            
            
            
                
                    100 μg | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $499.50
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
Description
CD34 Monoclonal specifically detects CD34 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| CD34 | |
| 1.0 mg/mL | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
| PBS with No Preservative | |
| 947 | |
| Partial recombinant human CD34 protein (aa32-290) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | 
| 756525 | |
| Flow Cytometry, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
| CD34 | |
| IgG2b | |
| Protein A or G purified | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title