Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD34 Antibody (756525), DyLight 550, R&D Systems™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB72272R
Description
CD34 Monoclonal specifically detects CD34 in Human samples. It is validated for Flow Cytometry.Specifications
CD34 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD34 | |
Partial recombinant human CD34 protein (aa32-290) | |
100 μL | |
Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
947 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
756525 | |
Flow Cytometry | |
CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction