Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD34 Antibody (HPCA1/1171), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247911PECY7
Description
CD34 Monoclonal specifically detects CD34 in Human, Rat samples. It is validated for Flow Cytometry, Product Image.Specifications
CD34 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
CD34 | |
Recombinant full-length human CD34 protein (Uniprot: P28906) | |
0.1 mL | |
Primary | |
This antibody recognizes a carbohydrate epitope on a single chain, transmembrane, heavily glycosylated protein of 90-120kDa, which is identified as CD34 (VI international workshop on human differentiation antigens). Its expression is a hallmark for identifying pluripotent hematopoietic stem or progenitor cells. Its expression is gradually lost as lineage committed progenitors differentiate. CD34 is a marker of choice for staining blasts in acute myeloid leukemia. In addition, it is expressed by soft tissue tumors, such as solitary fibrous tumor and gastrointestinal stromal tumor. CD34 expression is also found in vascular endothelium. Additionally, proliferating endothelial cells overexpress this molecule than the non-proliferating endothelial cells. Anti-CD34 labels 85% of angiosarcoma and Kaposis sarcoma, but shows low specificity. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
HPCA1/1171 | |
Flow Cytometry | |
CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
Mouse | |
Protein A or G purified | |
Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
947.0 | |
Human, Rat | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction