Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD34 Antibody (My10) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP2625540.025MG
Description
CD34 Monoclonal specifically detects CD34 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation.Specifications
CD34 | |
Monoclonal | |
Unconjugated | |
PBS | |
CD34 | |
KG-1a Human myeloblastic leukemic cell line (immature human myeloid cell line) from a patient with nonlymphocytic leukemia | |
947 | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
My10 | |
Western Blot, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
Rabbit | |
0.025 mg | |
Human CD34, an ∼105 - 120 kDa type 1 transmembrane glycoprotein expressed on the surface of most human hematopoietic stem and progenitor cells (HSPCs) as well as on mesenchymal stem cells, embryonic fibroblasts, endothelial cells, neurons, and some tumor cell lines. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction