Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 Antibody (QBEnd/10 + HPCA1/763) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$661.00

Specifications

Antigen CD34
Clone QBEnd/10 + HPCA1/763
Concentration 1.0 mg/mL
Dilution Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.25 - 0.5 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP247909A
SDP
View Documents
Novus Biologicals
NBP2479090.1MG
0.1 mg
Each for $661.00
Only null left
Add to Cart
 
NBP247909B
SDP
View Documents
Novus Biologicals
NBP2479090.2MG
0.2 mg N/A N/A N/A
Description

Description

CD34 Monoclonal specifically detects CD34 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Proximity Ligation Assay, Immunofluorescence, CyTOF-ready.
Specifications

Specifications

CD34
1.0 mg/mL
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Unconjugated
Mouse
Adaptive Immunity, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors
PBS with No Preservative
947
Detergent solubilized vesicular suspension prepared from human term placenta (QBEnd/10); Recombinant full-length human HPCA1 protein (HPCA1/763)
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
QBEnd/10 + HPCA1/763
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.25 - 0.5 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
Human
CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule
CD34
IgG
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.