Learn More
Description
Host Species: Mouse
Clone: QBEnd/10
Isotype: IgG1
Species Reactivity: Human. Does not react with dog and rat. Others not known
Immunogen: Detergent solubilized vesicular suspension prepared from a perfusate of human term placenta
Molecular Weight: 105-120 kDa
Positive Control: Tonsil
Cellular Localization: Membrane/cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | CD34 (Endothelial Cell Marker) Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | QBEnd/10 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Detergent solubilized vesicular suspension prepared from a perfusate of human term placenta |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.