Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 (Clone QBEnd/10) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS363R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
7 mL
Conjugate:
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
200μg/mL
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS363R1 1 mL Unconjugated Prediluted Prediluted
MS363P 1 mL Unconjugated Concentrated 200μg/mL
MS363R7 7 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS363R1 Supplier Epredia Supplier No. MS363R1

Obtain accurate, reproducible results in immunohistochemistry procedures with Epredia™ CD34 (Endothelial Cell Marker) Ab-1, Mouse Monoclonal Antibody.

CD34, a single chain transmembrane glycoprotein, is selectively expressed on human lymphoid and myeloid hematopoietic progenitor cells. Staining for CD34 has been used to measure angiogenesis, which reportedly predicts tumor recurrence. Antibody to CD34 also reacts with vascular endothelial cells in normal tissues and in benign and malignant proliferations. This antibody is of value in the study of benign and malignant vascular tumors as well as characterization of leukemias.

Host Species: Mouse

Clone: QBEnd/10

Isotype: IgG1

Species Reactivity: Human. Does not react with dog and rat. Others not known

Immunogen: Detergent solubilized vesicular suspension prepared from a perfusate of human term placenta

Molecular Weight: 105-120 kDa

Positive Control: Tonsil

Cellular Localization: Membrane/cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD34 (Endothelial Cell Marker) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone QBEnd/10
Concentration Prediluted
Conjugate Unconjugated
Formulation Ready-to-use for IHC
Host Species Mouse
Immunogen Detergent solubilized vesicular suspension prepared from a perfusate of human term placenta
Quantity 1 mL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.