Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD34 Monoclonal Antibody (4H11[APG]), PE
GREENER_CHOICE

Catalog No. 01676372
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-372 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-372 Supplier Invitrogen™ Supplier No. MA119645
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with extracellular class III epitope on CD34, a 110-115 kDa monomeric transmembrane phosphoglycoprotein expressed on hematopoietic progenitors cells and on the most pluripotential stem cells; it is gradually lost on progenitor cells. The antibody 4H11[APG] completely blocks binding of class III antibodies BIRMA K3 and 8G12 on KG1a cell line.

CD34 is a highly glycosylated monomeric with a molecular weight range of 111-115 kDa surface protein that is present on many stem cell populations. CD34 is a stem cell marker although its expression on human hematopoietic stem cells is reversible. CD34 may serve as a surface receptor that undergoes receptor-mediated endocytosis and regulates adhesion, differentiation and proliferation of hematopoietic stem cells and other progenitors. CD34 expression is likely to represent a specific state of hematopoietic development that may have altered adhering properties with expanding and differentiating capabilities in both in vitro and in vivo conditions. CD34 is possibly an adhesion molecule with a putative role for mediating the attachment of stem cells to the bone marrow extracellular matrix or directly to stromal cells. Further, CD34 could act as a scaffold for the attachment of lineage specific glycans, allowing stem cells to bind to lectins expressed by stromal cells or other marrow components. CD34 is thought to have a role in presenting carbohydrate ligands to selectins. The intracellular chain of the CD34 antigen is a site of phosphorylation by activated protein kinase C suggesting a putative role in signal transduction. Diseases associated with CD34 dysfunction include dermatofibrosarcoma and neurofibroma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD34
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 4H11[APG]
Concentration 0.06 mg/mL
Conjugate PE
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene Cd34
Gene Accession No. P28906
Gene Alias (E3-independent) E2 ubiquitin-conjugating enzyme R1; AU040960; CD34; CD34 antigen; CD34 molecule; CDC34; cell division cycle 34; cell division cycle 34 homolog; cluster designation 34; cluster of differentiation antigen 34; E2 ubiquitin-conjugating enzyme R1; E2-CDC34; hematopoietic progenitor cell antigen CD34; hematopoietic progenitor cell marker CD34; Mucosialin; RP11-328D5.2; sialomucin protein; type 1 transmembrane protein; UBC3; UBCH3; UBE2R1; ubiquitin carrier protein; ubiquitin conjugating enzyme E2 R1; ubiquitin-conjugating enzyme E2 R1; Ubiquitin-conjugating enzyme E2-32 kDa complementing; ubiquitin-conjugating enzyme E2-CDC34; Ubiquitin-protein ligase R1
Gene Symbols Cd34
Host Species Mouse
Immunogen Permanent human cell line from a patient with myeloid leukaemia.
Purification Method Size-exclusion chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 947
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.