Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD34 Mouse anti-Human, BV421, Clone: QBEND/10.rMAb (also known as QB-END-10, QBEND10, QBEND/10, QBEnd10), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 568861
Description
CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Flow cytometry
Specifications
CD34 | |
Monoclonal | |
Brilliant Violet 421 | |
CD34; CD34 antigen | |
Human Placental Vesicles | |
25 Tests | |
Primary | |
Human | |
Liquid |
Flow Cytometry | |
QBEND/10.rMAb (also known as QB-END-10, QBEND10, QBEND/10, QBEnd10) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
947 | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 λ2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction