Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD34 Mouse anti-Human, BV605, Clone: 8G12 (also known as HPCA2), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 745247
Description
CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Flow Cytometry.
Specifications
CD34 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
8G12 (also known as HPCA2) | |
Brilliant Violet 605 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction