Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 Mouse anti-Human, Clone: SPM610, Abnova™

Catalog No. 89933789 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-789 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-789 Supplier Abnova Corporation Supplier No. MAB13188
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant human CD34.

CD34 is a monomeric cell surface antigen with a molecular mass of approximately 110 kD that is selectively expressed on human hematopoietic progenitor cells.[supplied by OMIM]

Specifications

Antigen CD34
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SPM610
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human CD34.
Dilution Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determ
Formulation In 10mM PBS.
Gene Symbols CD34
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human CD34.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 947
Target Species Human
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.