Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 Mouse anti-Human, FITC, Clone: 581, BD

Catalog No. BDB560942 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560942 25 Tests
BDB555821 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560942 Supplier BD Biosciences Supplier No. 560942
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 581 monoclonal antibody specifically binds to CD34, a sialomucin-like type I transmembrane glycoprotein. This single-chain, 105-120 kDa, heavily O-glycosylated protein is expressed on hematopoietic progenitor cells, vascular endothelium, bone marrow stromal cells and embryonic fibroblasts. The cytoplasmic region of the CD34 antigen is a target for phosphorylation by activated protein kinase C suggesting CD34 may play a role in signal transduction. CD34 may also play a role as an adhesion molecule since it binds to CD62E and CD62L. Clone 581 binds to the class III CD34 epitope, it is resistant to neuraminidase, chymopapain and glycoprotease. The 581 antibody blocks reactivity of another anti-CD34 monoclonal antibody, 8G12.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD34
Applications Flow Cytometry
Classification Monoclonal
Clone 581
Conjugate FITC
Description gp105-120; My10; Hematopoietic progenitor cell antigen CD34
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.