Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD34 Mouse anti-Human, Oligo, Clone: 8G12 (also known as HPCA2), BD™ AbSeq ™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 940264
This item is not returnable.
View return policy
Description
CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Single Cell 3' Sequencing
Specifications
CD34 | |
Monoclonal | |
Oligo | |
CD34; CD34 antigen; CD34 molecule | |
Human KG-1a Cell Line | |
RUO | |
Human | |
IgG1 κ |
Single Cell 3' Sequencing | |
Clone 8G12 (also known as HPCA2) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction