Learn More
CD34 Mouse, anti-Human, R718, Clone: 8G12, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 752176
Description
The 8G12 monoclonal antibody specifically recognizes CD34, a 105-120 kDa single-chain type I transmembrane glycoprotein. The 8G12 antibody recognizes an epitope on CD34 distinct from the one recognized by clone My10. CD34 is expressed on immature hematopoietic precursor cells and all hematopoietic colony-forming cells in bone marrow and blood, including unipotent (CFU-GM, BFU-E) and pluripotent progenitors (CFU-GEMM, CFU-Mix, and CFUBlast). The CD34 antigen is a differentiation stage-specific leucocyte antigen. Terminal deoxynucleotidyl transferase-positive B- and T-lymphoid precursors in normal bone marrow are CD34+. The CD34 antigen is present on early myeloid cells that express the CD33 antigen but lack the CD14 and CD15 antigens and on early erythroid cells that express the CD71 antigen and dimly express the CD45 antigen. The CD34 antigen is also found on capillary endothelial cells and approximately 1% of human thymocytes. Normal peripheral blood lymphocytes, monocytes, granulocytes, and platelets do not express CD34. CD34 density is highest on early hematopoietic progenitor cells and decreases as cells mature. The antigen is absent on fully differentiated hematopoietic cells. Uncommitted CD34+ progenitor cells are CD38- and lack lineage-specific antigens such as CD71, CD33, CD10, and CD5, while CD34+ cells that are lineage-committed express the CD38 antigen in high density. Most CD34+ cells reciprocally express either the CD45RO or CD45RA antigens, with the CD45RO+ population being the more primitive. Approximately 60% of acute B-lymphoid leukemias and acute myeloid leukemias (AML) and 1% to 5% of acute T-lymphoid leukemias express CD34. CD34 is not expressed on chronic lymphoid leukemias or lymphomas.

Specifications
CD34 | |
Monoclonal | |
R718 | |
50 μg | |
Primary | |
Refrigerate Upon Arrival |
Flow Cytometry | |
8G12 | |
Mouse | |
RUO | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.