Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD34 Mouse anti-Human, RB545, BD OptiBuild™
Mouse Primary Antibody
Supplier: BD Biosciences 756399
Description
CD34 Primary antibody specifically detects CD34 in Human samples. It is validated for Flow Cytometry
Specifications
| CD34 | |
| 8G12 (also known as HPCA2) | |
| RB545 | |
| CD34; CD34 antigen; CD34 molecule | |
| Human KG-1a Cell Line | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| 947 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction