| Antigen |
CD34 |
| Applications |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification |
Monoclonal |
| Clone |
QBEnd/10 |
| Conjugate |
Unconjugated |
| Dilution |
Western Blot 0.25-0.5ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml |
| Formulation |
PBS with 0.05% BSA. with 0.05% Sodium Azide |
| Gene Accession No. |
P28906 |
| Gene Alias |
CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule |
| Gene Symbols |
CD34 |
| Host Species |
Mouse |
| Immunogen |
Detergent solubilized vesicular suspension prepared from human term placenta |
| Purification Method |
Protein A purified |
| Quantity |
0.02 mg |
| Regulatory Status |
RUO |
| Research Discipline |
Adaptive Immunity, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
947 |
| Test Specificity |
This MAb recognizes a single chain, transmembrane, heavily glycosylated protein of 90-120kDa, which is identified as CD34. On the basis of differential sensitivity to degradation by specific enzymes, epitopes of monoclonal antibodies to CD34 are classified into three main categories, class I, class II and class III. It is a class II antibody whose epitope is resistant to neuraminidase but sensitive to glycoprotease and chymopapain. CD34 expression is a hallmark for identifying pluripotent hematopoietic stem or progenitor cells. Its expression is gradually lost as lineage committed progenitors differentiate. CD34 is a marker of choice for staining blasts in acute myeloid leukemia. In addition, CD34 is expressed by soft tissue tumors, such as solitary fibrous tumor and gastrointestinal stromal tumor. Its expression is also found in vascular endothelium. Additionally, it appears that proliferating endothelial cells express this molecule more than the non-proliferating endothelial cells. Anti-CD34 labels > 85% of angiosarcoma and Kaposi s sarcoma, but with a lower specificity. |
| Target Species |
Human, Primate, Bovine (Negative), Canine (Negative), Rat (Negative), Sheep (Negative) |
| Content And Storage |
Store at 4C. |
| Form |
Purified |
| Isotype |
IgG1 κ |
|
Show More
Show Less
|
|