Learn More
Invitrogen™ CD34 Polyclonal Antibody

Description
Positive Samples: Mouse brain, Mouse testis, Rat brain; Cellular Location: Membrane, Single-pass type I membrane protein Immunogen sequence: SLDNNGTATP ELPTQGTFSN VSTNVSYQET TTPSTLGSTS LHPVSQHGNE ATTNITETTV KFTSTSVITS VYGNTNSSVQ SQTSVISTVF TTPANVSTPE TTLKPSLSPG NVSDLSTTST SLATSPTKPY TSSSPILSDI KAEIKCSGIR EVKLTQGICL EQNKTSSCAE FKKDRGEGLA RVLCGEEQAD ADAGAQVCSL LLAQSEVRPQ CLLLVLANRT EISSKLQLMK KHQSDLKKLG ILDFTEQDVA SHQSYSQKT.
Specifications
Specifications
| Antigen | CD34 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.86 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | Cd34 |
| Gene Accession No. | P28906, Q64314 |
| Gene Alias | (E3-independent) E2 ubiquitin-conjugating enzyme R1; AU040960; CD34; CD34 antigen; CD34 molecule; CDC34; cell division cycle 34; cell division cycle 34 homolog; cluster designation 34; cluster of differentiation antigen 34; E2 ubiquitin-conjugating enzyme R1; E2-CDC34; hematopoietic progenitor cell antigen CD34; hematopoietic progenitor cell marker CD34; Mucosialin; RP11-328D5.2; sialomucin protein; type 1 transmembrane protein; UBC3; UBCH3; UBE2R1; ubiquitin carrier protein; ubiquitin conjugating enzyme E2 R1; ubiquitin-conjugating enzyme E2 R1; Ubiquitin-conjugating enzyme E2-32 kDa complementing; ubiquitin-conjugating enzyme E2-CDC34; Ubiquitin-protein ligase R1 |
| Gene Symbols | Cd34 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.