Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD34, Rabbit anti-Human, Clone: Q19-E, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB15976
Specifications
CD34 | |
Monoclonal | |
Unconjugated | |
In 20 mM Tris-HCl buffer, pH 8.0 (20 mg/mL BSA, 0.05% Sodium Azide). | |
- | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody |
Immunohistochemistry (Paraffin) | |
Q19-E | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) The optimal working dilution should be determined by the end user. | |
CD34 molecule | |
CD34 | |
A synthetic peptide corresponding to N-terminus of human CD34. | |
RUO | |
947 | |
Store at 4°C. Do not freeze. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction