Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD34 Rat anti-Mouse, BV786, Clone: RAM34, BD Biosciences™
Rat Monoclonal Antibody
Supplier: BD Biosciences 742971
Description
CD34 Monoclonal antibody specifically detects CD34 in Mouse samples. It is validated for Flow Cytometry
Specifications
CD34 | |
Monoclonal | |
Brilliant Violet 786 | |
Cd34; CD34 antigen; Hematopoietic progenitor cell antigen CD34 | |
Recombinant Mouse CD34 | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
RAM34 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction