Learn More
Invitrogen™ CD34 (soluble) Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ 500P327BT500UG

Description
AA Sequence of recombinant protein: LDNNGTATPE LPTQGTFSNV STNVSYQETT TPSTLGSTSL HPVSQHGNEA TTNITETTVK FTSTSVITSV YGNTNSSVQS QTSVISTVFT TPANVSTPET TLKPSLSPGN VSDLSTTSTS LATSPTKPYT SSSPILSDIK AEIKCSGIRE VKLTQGICLE QNKTSSCAEF KKDRGEGLAR VLCGEEQADA DAGAQVCSLL LAQSEVRPQC LLLVLANRTE ISSKLQLMKK HQSDLKKLGI LDFTEQDVAS HQSYSQKT. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human sCD34. Anti-Human sCD34-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human sCD34 by sandwich ELISA (using 100 μL/well), a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human sCD34 (500-P327) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human sCD34. Western Blot: To detect Human sCD34 by Western Blot analysis, this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible development reagents, the detection limit for Recombinant Human sCD34 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P327BT-1MG will be provided as 2 x 500 μg
CD34 is a highly glycosylated monomeric with a molecular weight range of 111-115 kDa surface protein that is present on many stem cell populations. CD34 is a stem cell marker although its expression on human hematopoietic stem cells is reversible. CD34 may serve as a surface receptor that undergoes receptor-mediated endocytosis and regulates adhesion, differentiation and proliferation of hematopoietic stem cells and other progenitors. CD34 expression is likely to represent a specific state of hematopoietic development that may have altered adhering properties with expanding and differentiating capabilities in both in vitro and in vivo conditions. CD34 is possibly an adhesion molecule with a putative role for mediating the attachment of stem cells to the bone marrow extracellular matrix or directly to stromal cells. Further, CD34 could act as a scaffold for the attachment of lineage specific glycans, allowing stem cells to bind to lectins expressed by stromal cells or other marrow components. CD34 is thought to have a role in presenting carbohydrate ligands to selectins. The intracellular chain of the CD34 antigen is a site of phosphorylation by activated protein kinase C suggesting a putative role in signal transduction. Diseases associated with CD34 dysfunction include dermatofibrosarcoma and neurofibroma.
Specifications
CD34 (soluble) | |
Polyclonal | |
Biotin | |
Cd34 | |
(E3-independent) E2 ubiquitin-conjugating enzyme R1; AU040960; CD34; CD34 antigen; CD34 molecule; CDC34; cell division cycle 34; cell division cycle 34 homolog; cluster designation 34; cluster of differentiation antigen 34; E2 ubiquitin-conjugating enzyme R1; E2-CDC34; hematopoietic progenitor cell antigen CD34; hematopoietic progenitor cell marker CD34; Mucosialin; RP11-328D5.2; sialomucin protein; type 1 transmembrane protein; UBC3; UBCH3; UBE2R1; ubiquitin carrier protein; ubiquitin conjugating enzyme E2 R1; ubiquitin-conjugating enzyme E2 R1; Ubiquitin-conjugating enzyme E2-32 kDa complementing; ubiquitin-conjugating enzyme E2-CDC34; Ubiquitin-protein ligase R1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
947 | |
-20°C | |
Lyophilized |
ELISA, Western Blot | |
0.1-1.0 mg/mL | |
PBS with no preservative | |
P28906 | |
Cd34 | |
E.coli-derived, 27.4 kDa Recombinant Human sCD34. | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.