Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD344 Mouse, anti-Human, R718, Clone: CH3A4A7, BD OptiBuild™

Catalog No. BDB752194 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752194 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752194 Supplier BD Biosciences Supplier No. 752194
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CH3A4A7 monoclonal antibody reacts with an extracellular epitope on human CD344 which is also known as Frizzled family receptor 4 (FZD4) or Frizzled-4. CD344 is a member of the Frizzled family of proteins within the G protein-coupled receptor (GPCR) superfamily. CD344 is a receptor for Wnt proteins and participates in the activation of the Wnt/beta-catenin pathway (canonical Wnt signaling). Patients with familial exudative vitreoretinopathy (FEVR) carry mutations in Frizzled-4. This disease is characterized by retinal degeneration and hearing loss. On tissues, CD344 is almost ubiquitously expressed. CD344 is highly expressed in adult heart and skeletal muscle.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD344
Applications Flow Cytometry
Classification Monoclonal
Clone CH3A4A7
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.