Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD35 Antibody (7G6), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252667JF646
Description
CD35 Monoclonal specifically detects CD35 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD35 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CR1 | |
Purified Mouse CR1. | |
0.1 mL | |
1378 | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
7G6 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
complement component (3b/4b) receptor 1 (Knops blood group), KN | |
Rabbit | |
Protein A or G purified | |
Primary | |
Recognises an epitope shared by 145-150kDa and 190kDa complement receptor proteins, originally designated CR2 (CD21) and CR1 (CD35). | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction