Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD35 Antibody (7G6), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252667MFV500
Description
CD35 Monoclonal antibody specifically detects CD35 in Mouse samples. It is validated for Flow CytometrySpecifications
CD35 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
7G6 | |
Flow Cytometry | |
complement component (3b/4b) receptor 1 (Knops blood group), KN | |
Purified Mouse CR1. | |
0.1 mL | |
Immunology | |
1378 | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction