Learn More
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications
Specifications
| Antigen | CD35 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Clone | CR1/802 |
| Concentration | 0.2mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml |
| Formulation | 10mM PBS and 0.05% BSA with 0.05% Sodium Azide |
| Gene Accession No. | P17927 |
| Gene Alias | complement component (3b/4b) receptor 1 (Knops blood group), KN |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
