Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD35 Antibody (E11), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233080MFV450
Description
CD35 Monoclonal antibody specifically detects CD35 in Human, Monkey, Baboon, Cynomolgus Monkey, Macaque samples. It is validated for Flow CytometrySpecifications
CD35 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Monkey, Baboon, Cynomolgus Monkey, Macaque | |
Purified |
Flow Cytometry | |
E11 | |
Flow Cytometry | |
complement component (3b/4b) receptor 1 (Knops blood group), KN | |
Intact human monocytes | |
0.1 mL | |
Immunology | |
1378 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction