Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD35 Antibody (To5), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250177MFV500
Description
CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Flow CytometrySpecifications
CD35 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
To5 | |
Flow Cytometry | |
complement component (3b/4b) receptor 1 (Knops blood group), KN | |
C3b receptor-enriched precipitates from tonsil cell membranes | |
0.1 mL | |
Immunology | |
1378 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction