Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD35 Monoclonal Antibody (E11), Invitrogen™

Catalog No. PIMA513122
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513122 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513122 Supplier Thermo Scientific Supplier No. MA513122

Mouse Monoclonal Antibody

MA5-13122 targets CD35 in FACS, ICC/IF, and IHC (P) applications and shows reactivity with Human and Non-human primate samples. The MA5-13122 immunogen is intact human monocytes.

CD35 (CR1, Complement Receptor 1) is a cell membrane-bound, monomeric glycoprotein and its primary function is to serve as the cellular receptor for C3b and C4b, the most important components of the complement system leading to clearance of foreign macromolecules. CD35 protein mediates cellular binding to particles and immune complexes that have activated complement. The CD35 complex is four different allotypes (C,A,B,D) C is 160 kDa; A is 190 kDa; B is 220 kDa and D is 250 kDa. CD35 is involved in the processing of immune complexes, promoting of binding and phagocytosis of C3b complexes and inhibition of complement activation. CD35 is on neutrophils, eosinophils, monocytes, B-cells, some NK-cells, erythrocytes, myeloid leukemias, follicular dendritic reticulum cells and is negative on basophils. Decreases in expression of CD35 protein and/or mutations in its gene have been associated with gallbladder carcinomas, mesangiocapillary glomerulonephritis, systemic lupus erythematosus and sarcoidosis. Mutations in the CD35 gene have also been associated with a reduction in Plasmodium falciparum rosetting, conferring protection against severe malaria. Alternate allele-specific splice variants encoding different isoforms of CD35 have been characterized. An additional secreted form of CD35 have also been described but has not been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD35
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone E11
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene CR1
Gene Accession No. P17927
Gene Alias C3 binding protein; C3b/C4b receptor; C3-binding protein; C3BR; C4BR; CD35; CD35 antigen; complement component (3b/4b) receptor 1 (Knops blood group); Complement Component (3b/4b) receptor 1 including Knops blood group system; complement component 3b/4b receptor 1 (Knops blood group); complement receptor type 1; CR1; KN; Knops blood group antigen
Gene Symbols CR1
Host Species Mouse
Immunogen Intact human monocytes
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1378
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.