Learn More
Invitrogen™ CD35 Monoclonal Antibody (E11), PE

Description
CD35 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD35 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | E11 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with <0.1% sodium azide; pH 7.5 |
| Gene | CR1 |
| Gene Accession No. | P17927 |
| Gene Alias | C3 binding protein; C3b/C4b receptor; C3-binding protein; C3BR; C4BR; CD35; CD35 antigen; complement component (3b/4b) receptor 1 (Knops blood group); Complement Component (3b/4b) receptor 1 including Knops blood group system; complement component 3b/4b receptor 1 (Knops blood group); complement receptor type 1; CR1; KN; Knops blood group antigen |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.