Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD35 Mouse anti-Human, Brilliant Violet 510, Clone: E11, BD Optibuild™

Catalog No. BDB744275 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744275 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744275 Supplier BD Biosciences Supplier No. 744275
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The E11 monoclonal antibody specifically binds to CD35. CD35 is also known as Complement receptor type 1 (CR1), C3b/C4b receptor, C3BR, C4BR, Immune adherence receptor, or KN. CD35 is a type I transmembrane glycoprotein that exists in four allelic forms of 160, 190, 220 and 250 kDa. CD35 serves as a receptor for complement fragments C3b, iC3b, C3dg, C4b, iC3, and iC4. It enhances phagocytosis by neutrophils and monocytes and regulates complement activation. It is expressed on erythrocytes, granulocytes, monocytes, B cells, and some dendritic cells, T cells, and NK cells. It binds complement components C3b and C4b, mediating. This antibody cannot inhibit the phagocytic capacity of granulocytes. The CD35 antibody is useful in studies of cells that express complement receptors.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD35
Classification Monoclonal
Clone E11
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CR1, Complement receptor type 1, C3b/C4b receptor, C3BR, C4BR, KN
Host Species Mouse
Immunogen Human Cells of the Monocyte Lineage
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.