Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD35 Rat anti-Mouse, BUV615, Clone: 8C12, BD OptiBuild™

Catalog No. BDB752312 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752312 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752312 Supplier BD Biosciences Supplier No. 752312
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD35 Monoclonal antibody specifically detects CD35 in Mouse samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD35
Applications Flow Cytometry
Classification Monoclonal
Clone 8C12
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Ms CD35 BUV615 8C12 50 μg
Host Species Rat
Immunogen Purified mouse CR1
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD35
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2a κ
Reactivity Mouse
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.