Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD354 (TREM-1) Mouse anti-Human, BUV615, Clone: 6B1, BD OptiBuild™

Catalog No. BDB752319 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752319 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752319 Supplier BD Biosciences Supplier No. 752319
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD354 (TREM-1) Monoclonal antibody specifically detects CD354 (TREM-1) in Human samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD354 (TREM-1)
Applications Flow Cytometry
Classification Monoclonal
Clone 6B1
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Hu TREM-1 (CD354) BUV615 6B1 50 μg
Host Species Mouse
Immunogen Human Trem-1 Recombinant Protein
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD354 (TREM-1)
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG1 κ
Reactivity Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.