Learn More
CD355 (CRTAM) Monoclonal Antibody (Cr24.1), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$413.00
Specifications
Antigen | CD355 (CRTAM) |
---|---|
Clone | Cr24.1 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
53-355-942
![]() |
Invitrogen™
53355942 |
100 Tests |
Each of 1 for $413.00
|
|
|||||
Description
Description: This monoclonal antibody Cr24.1 reacts with human CD355, also known as Class-I MHC-restricted T-cell-associated molecule (CRTAM). CD355 usually exists as a homodimer, however, it forms a heterodimer with its ligand Necl-2. CD355 is transiently expressed by activated NK, NKT, gamma delta T, and CD8+ T cells. CD355 is also expressed by Purkinje neurons. Human CD355 is involved in immunosurveillance of cancer cells, gamma delta T cell survival, cell polarity, retention of lymphocytes in lymph nodes, promotion of cytotoxicity in NK cells, and IFN gamma secretion from CD8+ T cells. Cr24.1 has been reported to have antagonist activity. Applications Reported: This CR24.1 antibody has been reported for use by flow cytometric analysis. Applications Tested: This CR24.1 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser
The CRTAM gene is upregulated in CD4-positive and CD8-positive T cells and encodes a type I transmembrane protein with V and C1-like Ig domains.Specifications
CD355 (CRTAM) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
O95727 | |
CRTAM | |
Primary | |
CRTAM |
Cr24.1 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
56253 | |
IgG2a κ | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.