Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD355 (CRTAM) Monoclonal Antibody (Cr24.1), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 12355942
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-355-942 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-355-942 Supplier Invitrogen™ Supplier No. 12355942
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: This monoclonal antibody Cr24.1 reacts with human CD355, also known as Class-I MHC-restricted T-cell-associated molecule (CRTAM). CD355 usually exists as a homodimer, however, it forms a heterodimer with its ligand Necl-2. CD355 is transiently expressed by activated NK, NKT, gamma delta T, and CD8+ T cells. CD355 is also expressed by Purkinje neurons. Human CD355 is involved in immunosurveillance of cancer cells, gamma delta T cell survival, cell polarity, retention of lymphocytes in lymph nodes, promotion of cytotoxicity in NK cells, and IFN gamma secretion from CD8+ T cells. Cr24.1 has been reported to have antagonist activity. Applications Reported: This CR24.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CR24.1 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser

Class-I MHC-restricted T cell-associated molecule (CRTAM) is a gene located on chromosome 11q24.2, encoding a member of the immunoglobulin superfamily that plays a crucial role in immune cell interactions. CRTAM is expressed on activated CD8+ T cells, natural killer (NK) cells, and some subsets of CD4+ T cells, where it is involved in promoting cell adhesion, cytokine production, and cytotoxic responses. The protein binds to Necl-2 (CD112), mediating intercellular communication that influences immune responses, particularly in the context of viral infections and tumor surveillance. CRTAM's involvement in directing immune cell activity highlights its importance in the regulation of adaptive and innate immunity. Research into CRTAM is exploring its potential as a therapeutic target for modulating immune functions, aiming to enhance immune responses against cancer and infectious diseases or to attenuate autoimmune reactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD355 (CRTAM)
Applications Flow Cytometry
Classification Monoclonal
Clone Cr24.1
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CRTAM
Gene Accession No. O95727
Gene Alias CD355; CD355 antigen; class I MHC restricted T cell associated molecule; class I-restricted T cell-associated molecule; class-I MHC-restricted T cell associated molecule; class-I MHC-restricted T-cell-associated molecule; Crtam; cytotoxic and regulatory T cell molecule; cytotoxic and regulatory T-cell molecule
Gene Symbols CRTAM
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56253
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.