Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD355 (CRTAM) Rat anti-Mouse, RB780, Clone: 11-5, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 756459
Description
CD355 (CRTAM) Monoclonal antibody specifically detects CD355 (CRTAM) in Mouse samples. It is validated for Flow cytometry
Specifications
CD355 (CRTAM) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
11-5 | |
RB780 | |
class I-restricted T cell-associated molecule; crtam; cytotoxic and regulatory T-cell molecule | |
Mouse CRTAM extracellular sequence Recombinant Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction