Learn More
Invitrogen™ CD357 (AITR/GITR) Chimeric Recombinant Rat Monoclonal Antibody (DTA-1)
Rat Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA548162
Description
Specificity: This antibody is specific for murine GITR, a member of the tumor necrosis factor receptor (TNFR) family that is expressed at low levels on unstimulated T cells, B cells, and macrophages. Upon activation, CD4+ and CD8+ T cells up-regulate GITR expression, whereas immunoregulatory T cells constitutively express high levels of GITR. Preparation: This antibody was raised by immunising Wistar rats with the CD25+CD4+ T cell line. Hybridoma cells were generated by fusing spleen cells from the immunized rat with P3X63Ag8.653 myeloma cells, assessed for secreting antibodies capable of neutralizing in vitro CD25+CD4+ T cell-mediated suppression of anti-CD3-stimulated T cell proliferation9, and then cloned. This DTA-1 clone secreted a monoclonal rat immunoglobulin G2a (IgG2a) that showed high neutralizing activity.
Tumor necrosis factor receptor superfamily member 18 (TNFRSF18), also called GITR or AITR is a protein that in humans is encoded by the TNFRSF18 gene. It is mapped to 1p36.33. This gene encodes a member of the TNF-receptor superfamily. The encoded receptor has been shown to have increased expression upon T-cell activation, and it is thought to play a key role in dominant immunological self-tolerance maintained by CD25 (+)CD4 (+) regulatory T cells. Knockout studies in mice also suggest the role of this receptor is in the regulation of CD3-driven T-cell activation and programmed cell death. Three alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
Specifications
CD357 (AITR/GITR) Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
O35714 | |
Tnfrsf18 | |
This antibody was raised by immunising Wistar rats with the CD25+CD4+ T cell line. | |
200 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a λ |
Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot | |
DTA-1 | |
Unconjugated | |
Tnfrsf18 | |
Activation-inducible TNFR family receptor; AITR; CD357; GITR; GITR-D; glucocorticoid-induced TNFR-related protein; PRO364; TNF receptor superfamily activation-inducible protein; TNF receptor superfamily member 18; TNFRSF18; Tumor necrosis factor receptor superfamily member 18; tumor necrosis factor receptor superfamily, member 18; UNQ319; UNQ319/PRO364 | |
Rat | |
Protein A | |
RUO | |
21936 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.