Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD36 Antibody (1283D), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP254790R
Description
CD36 Monoclonal specifically detects CD36 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CD36/SR-B3 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD36 | |
Synthetic peptide made to an internal portion of the human CD36 protein (between amino acids 100-150) [UniProt P16671] | |
0.1 mL | |
Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers | |
948 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1283D | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction