Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD36 Antibody (1A7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2452500.1MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD36 Monoclonal specifically detects CD36 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional, Immunofluorescence.Specifications
CD36/SR-B3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
CD36 antigen, CD36 molecule (thrombospondin receptor), FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
Mouse | |
85 kDa | |
0.1 mg | |
Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers | |
948 | |
Human | |
Purified |
Flow Cytometry, SDS-Page, Immunofluorescence | |
1A7 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CD36 | |
Human CD36 from platelets | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a protein of 80kDa-90kDa, identified as CD36. It is expressed on platelets, monocytes and macrophages, microvascular endothelial cells, erythrocyte precursors, mammary epithelial cells, and some macrophage derived dendritic cells. CD36 acts as a receptor for thrombospondin (TSP), collagen types I, IV and V, P. falciparum malaria-infected erythrocytes, and sickle erythrocytes. It also functions as a scavenger receptor, mediating macrophage uptake of oxidized low-density lipoprotein (LDL) and recognition of apoptotic polymorphonuclear leukocytes (PMN). CD36 plays a role in platelet aggregation, macrophage foam cell development, inflammation, and the tissue ischemia observed in sickle cell disease and cerebral malaria. Note that 1-4% of Japanese and East Asia population lack CD36. | |
Store at 4C. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction