Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD36 Antibody (877302), Alexa Fluor 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB19553AF532
Description
CD36 Monoclonal specifically detects CD36 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry.Specifications
CD36/SR-B3 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CD36 | |
Partial recombinant human CD36 protein (amino acids 30-439) [UniProt P16671] | |
Primary | |
Human | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
877302 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
Mouse | |
0.1 mL | |
948 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction