Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD36 Antibody (GPIIIb/1654), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311597AF750
Description
CD36 Monoclonal antibody specifically detects CD36 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
CD36 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence | |
GPIIIb/1654 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
BDPLT10, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, leukocyte differentiation antigen CD36, PAS IV, PAS-4 protein, PASIV, platelet glycoprotein 4, platelet glycoprotein IV, SCARB3, scavenger receptor class B, member 3 | |
Recombinant human CD36 protein (Uniprot: P16671) | |
0.1 mL | |
Cancer, Cardiovascular Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lipid and Metabolism, Stem Cell Markers | |
948 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction