Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD36 Monoclonal Antibody (eBioNL07 (NL07)), PerCP-eFluor™ 710, eBioscience™

CD36 Monoclonal Antibody (eBioNL07 (NL07)), PerCP-eFluor 710, eBioscience™

Supplier:  LIFE TECHNOLOGIES 46036941

Encompass_Preferred

 View more versions of this product

Catalog No. 46-036-941


Only null left
Add to Cart

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD36
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P16671
CD36
Affinity chromatography
RUO
948
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
eBioNL07 (NL07)
PerCP-eFluor 710
CD36
adipocyte membrane protein; BDPLT10; Cd36; CD36 antigen; CD36 antigen (collagen type I receptor, thrombospondin receptor); CD36 molecule; CD36 molecule (thrombospondin receptor); CD36 protein; CHDS7; cluster determinant 36; collagen type I receptor thrombospondin receptor; collagen type I receptor, thrombospondin receptor; FAT; FAT/CD36; fatty acid translocase; fatty acid translocase/CD36; fatty acid transport protein; glycoprotein IIIb; GP3B; GP4; GPIIIB; GPIV; I79_011940; Leukocyte differentiation antigen CD36; PAS IV; PAS4; PAS-4; PAS-4 protein; PASIV; Platelet collagen receptor; Platelet glycoprotein 4; platelet glycoprotein IV; Scarb3; scavenger receptor class B, member 3; SR-B3; Thrombospondin receptor
Mouse
25 Tests
Primary
Human
Antibody
IgM
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.