Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD36 Mouse anti-Human, Biotin, Clone: OTI4H7, liquid, TrueMAB™
SDP

Catalog No. 501671303
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1303 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1303 Supplier OriGene Supplier No. TA400020
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD36 (fatty acid translocase, FAT) is an 88 kDa, ditopic glycosylated protein that belongs to the class B family of scavenger receptors. CD36 is expressed by most resting marginal zone B cells but not by follicular and B1 B cells, and is rapidly induced on Follicular B cells in vitro upon TLR and CD40 stimulation. CD36 does not affect the development of B cells, but modulates both primary and secondary antibody response. Similar to glucose transporter GLUT4, CD36 is translocated from intracellular pools to the plasma membrane following cell stimulation by insulin. In mouse, CD36 is responsible for gustatory perception of long-chain fatty acids. CD36 is preferentially found within lipid rafts, which facilitates its association with receptors, signaling and adapter molecules. CD36 binds to collagen, thrombospondin, anionic phospholipids and oxidized LDL. Further, CD36 may function as a cell adhesion molecule and directly mediates cyto-adherence of Plasmodium falciparum parasitized erythrocytes. Mutations in the CD36 gene cause platelet glycoprotein deficiency. Multiple alternatively spliced transcript variants encoding the CD36 protein have been found.

Specifications

Antigen CD36
Applications Western Blot
Classification Monoclonal
Clone OTI4H7
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide
Gene CD36
Gene Accession No. P16671
Gene Alias BDPLT10, CHDS7, FAT, GP3B, GP4, GPIV, PASIV, SCARB3
Gene Symbols CD36
Host Species Mouse
Immunogen Full length human recombit protein of human CD36 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 948
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.