Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD36 Mouse anti-Human, R718, Clone: CB38 (also known as NL07), BD Horizon™
Mouse Monoclonal Antibody
$164.70 - $339.50
Specifications
Antigen | CD36 |
---|---|
Clone | CB38 (also known as NL07) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Description
CD36 Monoclonal antibody specifically detects CD36 in Human samples. It is validated for Flow cytometry (Routinely Tested)Specifications
CD36 | |
Flow Cytometry | |
R718 | |
Mouse | |
Human | |
GPIIIb; Platelet GPIV; OKM5-antigen; PASIV | |
IgM κ | |
Affinity Chromatography |
CB38 (also known as NL07) | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Platelet Extracts | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title