Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD36 Mouse anti-Mouse, BB515, Clone: CRF D-2712, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 565933
Description
CD36 Monoclonal antibody specifically detects CD36 in Mouse samples. It is validated for Flow cytometry.
Specifications
CD36 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
25 μg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
CRF D-2712 | |
Brilliant Blue 515 (BB515) | |
Platelet glycoprotein 4; PAS-4; GPIIIB; Fatty acid translocase; Scavenger R | |
Recombinant mouse CD36 | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
IgA κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction