Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD36/SR-B3 Antibody (255606), DyLight 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB19551E
This item is not returnable.
View return policy
Description
CD36/SR-B3 Monoclonal antibody specifically detects CD36/SR-B3 in Human samples. It is validated for Flow CytometrySpecifications
| CD36/SR-B3 | |
| Monoclonal | |
| DyLight 405 | |
| BDPLT10, CD_antigen: CD36, CD36, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, PAS IV, PAS-4, PAS-4 protein, PASIV, Platelet collagen receptor, platelet glycoprotein 4, SCARB3, scavenger receptor class B, member 3, Thrombospondin receptor | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD36/SR-B3, Gly30-Asn439, Accession # P16671 | |
| 0.5 mL | |
| Cardiovascular Biology | |
| 948 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| 255606 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction