Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD36/SR-B3 Antibody (255606), PerCP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB19551C
Description
CD36/SR-B3 Monoclonal antibody specifically detects CD36/SR-B3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
CD36/SR-B3 | |
Monoclonal | |
PerCP | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
255606 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
BDPLT10, CD_antigen: CD36, CD36, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, PAS IV, PAS-4, PAS-4 protein, PASIV, Platelet collagen receptor, platelet glycoprotein 4, SCARB3, scavenger receptor class B, member 3, Thrombospondin receptor | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD36/SR-B3, Gly30-Asn439, Accession # P16671 | |
0.5 mL | |
Cardiovascular Biology | |
948 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction