Learn More
CD360 (IL-21 Receptor) Monoclonal Antibody (2SX21R), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$460.00
Specifications
Antigen | CD360 (IL-21 Receptor) |
---|---|
Clone | 2SX21R |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-113-0804
![]() |
Invitrogen™
46360142 |
100 Tests |
Each of 1 for $460.00
|
![]() |
|||||
Description
Description: This 2SX21R monoclonal antibody reacts with human CD360 (Interleukin-21 Receptor, IL-21R). IL-21R forms a heterodimeric receptor complex with the common gamma chain to form a functional receptor for IL-21. IL-21 is a class I cytokine of the common gamma chain family of cytokines and signals through the IL-21R complex to activate the JAK/STAT (Janus Kinase/Signal Transducers and Activators of Transcription) signaling pathway. Activation of the JAK/STAT pathway by IL-21 promotes transcription of genes responsive to STAT1 and STAT3. IL-21 is mainly produced by CD4+ T cells and mediates the proliferation, differentiation, and effector functions of cells expressing IL-21R such as B cells, T cells, NK cells, dendritic cells and other populations of myeloid cells. IL-21 and its receptor are implicated in the pathogenesis of chronic viral infections, autoimmunity, allergy, organ transplantation, and cancer. Applications Reported: This 2SX21R antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2SX21R antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
The protein encoded by this gene is a cytokine receptor for interleukin 21 (IL21). It belongs to the type I cytokine receptors, and has been shown to form a heterodimeric receptor complex with the common gamma-chain, a receptor subunit also shared by the receptors for interleukin 2 (IL2) and interleukin 5 (IL5). This receptor transduces the growth promoting signal of IL21, and is important for the proliferation and differentiation of T cells, B cells, and natural killer (NK) cells. The ligand binding of this receptor leads to the activation of multiple downstream signaling molecules, including JAK1, JAK3, STAT1, and STAT3. Knockout studies of a similar gene in mouse suggest a role for this gene in regulating immunoglobulin production. Three alternatively spliced transcript variants encoding the same protein have been described.Specifications
CD360 (IL-21 Receptor) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q9HBE5 | |
IgG1 κ | |
Affinity Chromatography | |
Antibody |
2SX21R | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
50615 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.