Learn More
Invitrogen™ CD365 (Tim-1) Monoclonal Antibody (RMT1-4), Biotin, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Supplier: Invitrogen™ 13586181

Description
Description: The RMT1-4 monoclonal antibody reacts with mouse CD365 (also known as TIM1 and HAVCR1) and is a member of the T cell immunoglobulin domain, mucin-like domain (TIM) gene family. CD365 has been found on activated CD4+ T cells, mast cells, and a subset of B cells. CD365 is involved in renal injury, tissue homeostasis, and T cell costimulation. CD365 promotes the differentiation of Th2 cells and the production of IL-4. CD365 can bind several ligands, including Hepatitis A virus, phosphatidylserine, TIM4, IgA, as well as itself. CD365 and TIM4 can mediate uptake of apoptotic cells through recognition of phosphatidylserine. CD365, as well as the other TIM family members, are associated with susceptibility to atopic and autoimmune diseases. In humans, certain polymorphic variants of CD365 are strongly associated with protection against atopy, and this association occurs only in individuals who have had past infection with hepatitis A virus (HAV). Applications Reported: This RMT1-4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RMT1-4 antibody has been tested by flow cytometric analysis of mouse CD365 (TIM1) transfected cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
CD365 also known as T-cell immunoglobulin and mucin domain 1 (Tim-1) is a type 1 surface glycoprotein that consists of extracellular N-terminal immunoglobulin V-like domain and a mucin domain, and a short cytoplasmic tail. The endogenous ligand for CD365 is Tim-4. In addition, CD365 is known to be a cellular receptor or co-receptor for a number of viruses including hepatitis A and C viruses, Ebola virus, Marburg virus, Dengue virus, Zika virus, West Nile virus, and Japanese encephalitis virus. The direct binding of the viruses by CD365 occurs via phosphatildyl-serine exposed on viral envelopes. Polymorphisms within human CD365 gene (HAVCR1) are associated with susceptibility to infection and the disease severity. CD365 has also been implicated in the T cell co-stimulation. CD365 is primarily expressed in epithelial cells. It is also highly expressed in A549, 769-P and Caco-2 and cell lines. CD365 exists also as a soluble protein generated by metalloproteinase mediated proteolytic cleavage of the surface protein.
Specifications
CD365 (Tim-1) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q5QNS5 | |
Havcr1 | |
Affinity Chromatography | |
RUO | |
171283 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
RMT1-4 | |
Biotin | |
Havcr1 | |
AI503787; CD365; HAVCR; HAVCR1; HAVcr-1; Hepatitis A virus cellular receptor 1; hepatitis A virus cellular receptor 1 homolog; Kidney injury molecule 1; Kim1; KIM-1; T cell immunoglobin domain and mucin domain protein 1; t cell immunoglobulin and mucin domain-containing protein 1; T cell membrane protein 1; T-cell immunoglobulin and mucin domain containing 1; T-cell immunoglobulin and mucin domain-containing protein 1; T-cell immunoglobulin mucin family member 1; T-cell immunoglobulin mucin receptor 1; T-cell membrane protein 1; TIM; TIM1; TIM-1; TIMD1; TIMD-1 | |
Rat | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.