Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD365 (TIM-1) Mouse anti-Human, Brilliant Violet 650, Clone: 1D12, BD Optibuild™

Catalog No. BDB742542 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742542 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742542 Supplier BD Biosciences Supplier No. 742542
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1D12 monoclonal antibody specifically binds to CD365, the T-cell immunoglobulin mucin receptor 1 (TIM-1). TIM-1 is expressed on kidney epithelial cells, T cells, and some hematopoietic and non-hematopoietic cells. CD365 (TIM-1) is a type 1 transmembrane glycoprotein that serves as a receptor for hepatitis A virus and is encoded by the HAVCR1 /(Hepatitis A virus cellular receptor 1) gene. TIM-1 also serves as a receptor for phosphatidylserine which is exposed on the surface of apoptotic cells. TIM-1 can reportedly mediate the uptake of apoptotic cells through the recognition of phosphatidylserine and thus help maintain tissue homeostasis and self-tolerance. TIM-1 is likewise known as Kidney injury molecule 1 (KIM-1). It is highly expressed by cancerous kidneys, and upregulated in the proximal tubular epithelium and shed into the urine during acute and chronic kidney injury. CD365 (TIM-1) also functions as a costimulatory molecule for immune cells. It is expressed by activated CD4+ T cells and regulates the effector functions (eg, enhanced cytokine production) and survival of differentiated T cells, including those mediating Th2-like immune responses. Other ligands have been described for TIM-1 including TIM-4 and LMIR5 (also known as CD300b) which are expressed by myeloid cells. With respect to disease associations, the HAVCR1 /gene has been linked to asthma, allergy, and some autoimmune diseases.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD365 (TIM-1)
Classification Monoclonal
Clone 1D12
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD365, TIM1, TIM, TIMD1, TIMD-1, HAVCR1, HAVCR-1, HAVCR, KIM1, KIM-1
Host Species Mouse
Immunogen Human TIM-1 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.