Learn More
Invitrogen™ CD365 (Tim-1) Recombinant Rabbit Monoclonal Antibody (P365D), Alexa Fluor™ 647, eBioscience™
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ 51365842

Description
Description: This P365D recombinant rabbit monoclonal antibody reacts with human CD365, which is also known as Tim-1, Kim-1, or Havcr1. Applications Tested: This P365D antibody has been pre-diluted and tested by flow cytometric analysis of 769P cells. This may be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser Filtration: 0.2 μm post-manufacturing filtered.
CD365 also known as T-cell immunoglobulin and mucin domain 1 (Tim-1) is a type 1 surface glycoprotein that consists of extracellular N-terminal immunoglobulin V-like domain and a mucin domain, and a short cytoplasmic tail. The endogenous ligand for CD365 is Tim-4. In addition, CD365 is known to be a cellular receptor or co-receptor for a number of viruses including hepatitis A and C viruses, Ebola virus, Marburg virus, Dengue virus, Zika virus, West Nile virus, and Japanese encephalitis virus. The direct binding of the viruses by CD365 occurs via phosphatildyl-serine exposed on viral envelopes. Polymorphisms within human CD365 gene (HAVCR1) are associated with susceptibility to infection and the disease severity. CD365 has also been implicated in the T cell co-stimulation. CD365 is primarily expressed in epithelial cells. It is also highly expressed in A549, 769-P and Caco-2 and cell lines. CD365 exists also as a soluble protein generated by metalloproteinase mediated proteolytic cleavage of the surface protein.
Specifications
CD365 (Tim-1) | |
Recombinant Monoclonal | |
5 μL/Test | |
PBS, proprietary buffer with 0.09% sodium azide; pH 7.2 | |
Q96D42 | |
Havcr1 | |
Protein corresponding to Human CD365 (aa22-aa129). | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry | |
P365D | |
Alexa Fluor 647 | |
Havcr1 | |
AI503787; CD365; HAVCR; HAVCR1; HAVcr-1; Hepatitis A virus cellular receptor 1; hepatitis A virus cellular receptor 1 homolog; Kidney injury molecule 1; Kim1; KIM-1; T cell immunoglobin domain and mucin domain protein 1; t cell immunoglobulin and mucin domain-containing protein 1; T cell membrane protein 1; T-cell immunoglobulin and mucin domain containing 1; T-cell immunoglobulin and mucin domain-containing protein 1; T-cell immunoglobulin mucin family member 1; T-cell immunoglobulin mucin receptor 1; T-cell membrane protein 1; TIM; TIM1; TIM-1; TIMD1; TIMD-1 | |
Rabbit | |
Protein A | |
RUO | |
26762 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid | |
eBioscience™ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.