Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD366 (TIM-3) Rat anti-Mouse, Alexa Fluor™ 647, Clone: RMT3-23, BD Phrmingen™
Rat Monoclonal Antibody
$40.00 - $100.40
Specifications
Antigen | CD366 (TIM-3) |
---|---|
Clone | RMT3-23 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD366 (TIM-3) Monoclonal antibody specifically detects CD366 (TIM-3) in Mouse samples. It is validated for Flow cytometrySpecifications
CD366 (TIM-3) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
171285 | |
IgG2a κ | |
Affinity Chromatography |
RMT3-23 | |
Flow Cytometry | |
Alexa Fluor 647 | |
Rat | |
Mouse | |
Cd366; TIM-3; Tim3; TIMD-3; Timd3; HAVcr-2; Havcr2 | |
Mouse TIM-3 Recombinant Protein | |
Primary | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title